Fowler 52-265-006 6in/150MM Rigid Pre-Cert. Rule

Representative image only.

Fowler 52-265-006 6in/150MM Rigid Pre-Cert. Rule

Fowler | Mfr: 522650060| Item: 9760457

Certified Rulers and Sets

₹13,700.00INR / Each

for an additional
₹7,200.00INR / Each
for an additional
₹19,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹13,700.00INR / Each

Buy It Together

Current Item

Fowler 52-265-006 6in/150MM Rigid Pre-Cert. Rule
Fowler 52-265-006 6in/150MM Rigid Pre-Cert. Rule
9760457

₹13,700.00INR / Each
+
InnoCal NIST-Traceable Calibration; Ruler, 0-60"
1705016

₹7,200.00INR / Each
=
Total Price 20900.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

6"/150mm rigid ruler
Graduations in 32nds, 64ths, 1mm, and 0.5mm
Includes NIST-traceable calibration report

1 Year Warranty
Specifications
  • Description6in/150MM Rigid Pre-Cert. Rule
  • Warranty1 year
Reviews
Q&A