Fluke VPS41 Voltage Probe Set, 40 MHz, CAT IV 600 V

Representative image only.

Fluke VPS41 Voltage Probe Set, 40 MHz, CAT IV 600 V

Fluke | Mfr: VPS41| Item: 2000016

Upgrade and expand your capabilities

₹68,300.00INR / Each

₹68,300.00INR / Each
Product Description

Key Features

  • Choose among universally compatible and model-specific items
  • Voltage probe set measures up to 40 MHz
  • 1 year warranty

More About this Item

Find test leads, adapters, and much more!

1 Year Warranty
Specifications
  • Compatible WithFluke Oscilloscopes
  • DescriptionVoltage Probe Set, 40 MHz, CAT IV 600 V
  • Warranty1 year
Reviews
Q&A