Fluke MODULIFT2L Ladder-Pulley System, 80-ft Rope
Representative image only.
Fluke MODULIFT2L Ladder-Pulley System, 80-ft Rope
Fluke | Mfr: FLUKE-MODULIFT2L| Item: 1778506
Make solar panel installation faster, safer, and more efficient!
₹541,000.00INR / Each
₹541,000.00INR / Each
Product Description
Includes
Pulley system, module handle assembly, three ramps, a 60-ft rope, and a duffle bag.
Key Features
  • Compatible with Werner or Louisville fiberglass extension ladder (not included)
  • Compact and simple to operate
  • Meet OSHA ladder safety standards
  • Reduce the risk of damaging the panels and the roof
  • Patented module “hook” securely holds panels for easy lifting
  • A brake prevents the module from sliding back down
  • Comes with an 80-ft rope
  • 1 year warranty
More About this Item
Safely and easily lift solar panels to the roof with the ladder-pulley system. This compact and simple-to-operate module lift system eliminates the need for heavy lifting as well as minimizes damaging the panels and the roof. Ladder set up takes less than 5 minutes by one person.
To operate, simply attach the edge of a PV module frame to a patented module “hook” to prevent any lateral sliding. When the rope is pulled to raise the module, it slides along the outward facing surface of the ladder rails. The included ramps provide continuities in the ladder surface for smooth transition between sections in the ladder. In the event the operator releases the rope too soon, a brake prevents the module from accidentally sliding down.
1 Year Warranty
Specifications
  • Compatible WithWerner or Louisville fiberglass extension ladder
  • DescriptionPV Module Lift™ Ladder-Pulley System, 80-ft Rope