Fluke Laser Shaft Alignment Tool
Representative image only.
Fluke Laser Shaft Alignment Tool
Fluke | Mfr: FLUKE-831| Item: 2005134
With its rugged sensALIGN® 3 sensor and reflector, this laser alignment tool can handle almost any mainstream machine alignment challenge
₹3,766,000.00INR / Each
₹3,766,000.00INR / Each
Product Description
Includes
Sensor laser, reflector prism, chain-type mounting bracket with 150 mm support posts (2x), 300 mm support posts (4x), lens cleaning cloth, USB sensor cable, USB PC cable, USB drive, USB drive cable, tape measure, power adapter (2x), quick reference guide, safety information sheet, and carrying case.
Key Features
  • Fast, easy, and precise alignment that adapts to your needs
  • Offers the highest-in-class IP rating
  • Single-laser technology for the fastest, most accurate laser alignment
  • Cloud connection enables real-time collaboration
  • Use up to 8 measurement points to achieve precision alignment on vertical machines
  • 2 year warranty
More About this Item
The Fluke 831 has an intuitive guided user interface that enables quick and complete shaft alignment without advanced training or complicated programs. It leverages powerful tools like unique extend mode to handle gross misalignment and integrated thermal growth calculator that automatically factors the dynamic machine changes into the result. Its quickly mounted setup and tablet-like interface make the 831 more user-friendly than any other conventional measurement methods. Align coupled shafts in four quick, easy steps. The user can leverage its integrated Wi-Fi cloud solution to easily transfer data from the 831 handheld device to the ARC 4.0 PC software.
Two year warranty
Specifications
  • Battery1 3.8V Lithium-ion rechargeable (included)
  • DescriptionLaser Shaft Alignment Tool
  • Warranty2 years