Fluke IRR1-SOL Solar Irradience Meter with Temperature Probe and Carry Case
Representative image only.
Fluke IRR1-SOL Solar Irradience Meter with Temperature Probe and Carry Case
Fluke | Mfr: FLK-IRR1-SOL| Item: 2003741
Surveying, installing, executing maintenance or checking performance of solar panels or a photovoltaic system, the Fluke IRR1-SOL Irradiance Meter gives you the 4 critical measurements needed all in one easy-to-use tool
₹143,000.00INR / Each
₹143,000.00INR / Each
Product Description
Key Features
  • Measure solar irradiance, ambient and PV module temperature, array orientation and tilt angles
  • Make instantaneous measurements to determine the watts per square meter solar irradiation, required by IEC 62446-1 standard
  • High contrast LCD with large numbers for easy readability in direct sunlight
  • Includes convenient carry case with shoulder strap
  • 3 year warranty
More About this Item
The Fluke IRR1-SOL Irradiance Meter has been designed from the ground up to simplify the installation, commissioning, and troubleshooting of photovoltaic arrays, measuring irradiance, temperature, inclination and direction of the solar array in a single handheld tool. With a rugged, compact design, a protective carrying case, and an easy-to-read, high-contrast LCD screen to read measurements in direct sunlight, the IRR1-SOL can go where you go. The simple user interface, instantaneous solar irradiation measurements and built-in temperature sensor make it easy to meet the IEC 62446-1 requirements for testing, documenting, and maintaining photovoltaic systems. Additionally, the integrated compass and inclination sensor allow you to quickly measure and document roof and site orientation, pitch, and panel tilt while surveying, installing, or adjusting an installation.
3 Year Warranty
Specifications
  • Intensity Range0 to 1400 W/m²
  • Intensity Resolution1 W/m²
  • Power4 AA Alkaline batteries
  • DescriptionSolar Irradience Meter
  • Warranty3 Years