Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Insulation Multimeter Kit

Representative image only.

Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Insulation Multimeter Kit

Fluke | Mfr: FLUKE-SOL-INS37-KIT| Item: 1778510

Optimize solar panel performance with high-efficiency PV analyzer I-V curve tracer kits

₹7,323,000.00INR / Each

₹7,323,000.00INR / Each
Product Description

Includes

PVA-1500HE2 analyzer: SolSensor sensor, clamp, MC4 wrenches, test lead sets (banana to alligator clip and banana to MC4), two type K thermocouple sensors, 50 adhesive discs for thermocouple attachment, sensor cleaning supplies, charging supplies, soft case, and hard carrying case.
1587 FC insulation tester: test leads, alligator clips, remote probe, type K thermocouple probe, and soft case.

Key Features

  • Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays
  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Pinpoint problems early to maximize solar energy production while minimizing downtime
  • Test up to 1500 V and up to 30 A
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
  • Kit includes a PVA-1500HE2 analyzer and 1587 FC insulation multimeter
  • Insulation multimeter with test voltage up to 1000 V and resistance measurements up to 2 GO
  • Pi/DAR timed ratio tests with TrendIt™ graphs identifies problems faster
  • Memory storage through Fluke Connect® App
  • Live circuit detection prevents insulation test if voltage of >30 V is detected
  • 1 year warranty

More About this Item

Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PV analyzer kits. Select from four kits that meet your application needs. Each kit includes a PVA-1500HE2 analyzer I-V curve tracers and additional Fluke meter(s) to help maximize efficiency and optimize PV performance. Download the data sheet for each instrument for complete specifications.

The high-efficiency PV analyzer is ideal for testing high-efficiency modules (>19% module efficiency). The high-efficiency modules have a large in-rush current when you start the test that can overheat the test instrument and slow down testing. Uniquely designed high-efficiency PV analyzer measures all string types, including high-efficiency modules, up to 30 A. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules.

Measure irradiance and module temperature by clamping the wireless SolSensor™ PV reference sensor (included with analyzer) to the module frame. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.

A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.

Help detect potential safety hazards and prevent system failures using the insulation multimeter. The multimeter provides insulation resistance testing up to 1000 V and resistance measurements up to 2 GO. This digital insulation tester and true-rms digital multimeter combination offers maximum versatility for both troubleshooting and preventative maintenance, The full-featured handheld insulation multimeter works with the PV analyzer to offer comprehensive diagnostics for solar arrays.

Conformite Europeenne1 Year Warranty
Specifications
  • DC Min (Volts)20
  • DC Max (Volts)1500
  • DC Voltage Accuracy±(0.5% ±0.25V)
  • DC Max (amps)30
  • DC Current Accuracy±(0.5% ±0.04A)
  • Display1537: large LCD
  • Memory1537: 99 insulation tests
  • InterfacePVA-1500HE2: Wi-Fi
    1537: USB
  • Safety RatingPVA-1500HE2: CAT III 1500 V; IEC 61010-1: Pollution Degree 2
    1537: IEC 61010-1, Pollution Degree 2, IEC 61557-1; IEC 61010-2-030: CAT IV 600 VIEC 61010-2-034: 2500 V DC
  • PowerPVA-1500HE2 and SolSensor: rechargeable batteries
    1537: 8 AA batteries
  • DescriptionHigh-Efficiency PV Analyzer I-V Curve Tracer and Insulation Tester Kit
  • Warranty1 year
Reviews
Q&A