Fluke 902 FC True-RMS HVAC Clamp Meter
Representative image only.
Fluke 902 FC True-RMS HVAC Clamp Meter
Fluke | Mfr: FLUKE-902 FC| Item: 2000001
Remote viewing of streaming data keeps you safe
₹133,000.00INR / Each
for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹133,000.00INR / Each
Buy It Together
Current Item
Fluke 902 FC True-RMS HVAC Clamp Meter
Fluke 902 FC True-RMS HVAC Clamp Meter
2000001
₹133,000.00INR / Each
+ =
Total Price 161000.00 ADD 2 ITEMS TO CART
Product Description
Includes
test leads, temperature probe, soft carrying case, and two AA alkaline batteries.
Key Features
  • Remain outside the arc flash zone while monitoring HVAC changes and troubleshooting
  • App is compatible with Apple® and Android™ devices
  • Measures flue gas temperature, start and run motor capacitors, and conducts flame rod testing
More About this Item
This true-rms clamp meter performs all of the functions typical for HVAC maintenance and troubleshooting, but with key improvements over the previous meter (model 902). The new meter features an expanded resistance range and a more ergonomic design, which is compatible with the convenient magnetic strap hanging kit (order separately). The most helpful upgrade to the new meter is the Fluke Connect interface, which communicates with Bluetooth®*-enabled devices to live-stream data, graph trends, and create and send reports.
*Note: The Bluetooth® wordmark and logos are registered trademarks owned by Bluetooth SIG, Inc.
3 Year Warranty
Specifications
  • DescriptionTrue-RMS HVAC Clamp Meter