Fluke 835 Laser Belt Alignment Tool with Reflector
Representative image only.
Fluke 835 Laser Belt Alignment Tool with Reflector
Fluke | Mfr: 5255306| Item: 3002828
Get more out of your belt driven systems through proper alignment
₹610,000.00INR / Each
₹610,000.00INR / Each
Product Description
Includes
Fluke 835 laser unit (green laser), 4 AAA batteries, Fluke 835 reflector unit, fabric carrying pouch, safety info, and Quick Reference Guide.
Key Features
  • One person operation – no training required
  • Reflected laser technology doubles the distance, enhancing accuracy
  • Light, robust, rugged tool you can take anywhere
  • Strong magnets mount to any size pulley or sprocket
  • 2 year warranty
More About this Item
The Fluke 835 laser belt alignment tool is great for precision-alignment belt-drive machines in your facility. Pulley alignment reduces belt wear, power, losses, and vibration of machinery, improving the reliability of your equipment. The green laser is brighter and more visible than traditional red lasers, especially on gray, which results in better data accuracy. It simultaneously shows offset, vertical, and horizonal angles, saving you time as cross-checking is not required.
Two year warranty
Specifications
  • Min operating temperature (° F)104
  • Max operating temperature (° F)23
  • Min operating temperature (° C)40
  • Max operating temperature (° C)-5
  • Wavelength505 - 535 nm (green – more visible)
  • Class2
  • Output< 1.0 mW
  • Battery Type4 AAA (included)
  • DescriptionLaser Belt Alignment Tool with Reflector
  • Warranty2 Years