Fluke 2000ACC Test Lead Accessory Kit for 2052/2062 Advanced Wire Tracers
Representative image only.
Fluke 2000ACC Test Lead Accessory Kit for 2052/2062 Advanced Wire Tracers
Fluke | Mfr: FLUKE-2000ACC| Item: 1778460
Ensure flexibility when tracing wires with a range of adapters and connectors
₹48,500.00INR / Each
₹48,500.00INR / Each
Product Description
Includes
Two 1 m test leads (red, black), one 7 m test lead (green), two test probes (black), two alligator clips (red, black), two outlet blade adapters (red, black), and two outlet round adapters (red, black).
Key Features
  • 7 m (23 ft) long green test lead helps with grounding away from the circuit being traced
  • Black stackable test lead allows stacking for convenience
  • Flexible springs and long test probes are ideal for hard-to-reach areas
  • 3 year warranty
More About this Item
The test lead accessory kit comes with adapters and connectors to help you trace energized and de-energized wires. Use the outlet round and blade adapters for connecting test leads to outlets; directly connect to bare conductors with alligator clips. Test probes fit securely between 4 and 8 mm sockets.
Measurement category:
CAT IV V (test leads and alligator clips); CAT II 1000 V (test probes); CAT II 300 V (outlet adapters)
Operating voltage and current:
600 V, 10 A max (red/black leads, green lead, and alligator clips); 1000 V, 8 A max (black probe); 300 V, 10 A max (outlet adapters)
3 Year Warranty
Specifications
  • Compatible WithFluke Advanced Wire Tracers 17782-72 and -83
  • DescriptionFluke Test Lead Accessory Kit for Fluke 2052/2062 Advanced Wire Tracers
  • Warranty3 years