Fluke 154 HART Calibration Assistant
Representative image only.
Fluke 154 HART Calibration Assistant
Fluke | Mfr: Fluke-154 US/CAN| Item: 1610138
Stay out of harm's way—wireless communication up to 250 ft (76 m)
Call For Price
Call For Price
Product Description
Includes
tablet, HART modem, hook test clips, alligator clips, USB tablet charger, USB modem charger, magnetic hanging strap, carrying case, and rechargeable batteries.
Key Features
  • Communication via Bluetooth®* enables remote calibration and device configuration
  • Programmed with different versions of HART to ensure compatibility with your device
  • Standalone or combine with a HART-enabled process calibrator
More About this Item
The HART calibration assistant makes device configuration and calibration safe and easy. Just connect the wireless modem's cable directly to the HART transmitter device or combine with a HART-enabled process calibrator. The FlukeHART mobile app installed on the tablet provides an intuitive interface for remotely controlling your device.
For sale in the United States and Canada only.
*The Bluetooth® word mark and logos are registered trademarks owned by Bluetooth SIG, Inc.
3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Display TypeTablet operating system: Android™ 5.0 or newer
  • OutputHART interface: HART 4, 5, 6, and 7
  • ConnectionsBluetooth® (modem) and Wi-Fi (Internet)
  • BatteriesRechargeable Li-ion
  • Battery life19 hrs continuous use
  • DescriptionHART Calibration Assistant