Flir MR06 Wall Cavity Probe for MR-Series Moisture Meters
Representative image only.
Flir MR06 Wall Cavity Probe for MR-Series Moisture Meters
Flir | Mfr: MR06| Item: 3780214
Update your moisture meter
₹70,600.00INR / Each
₹70,600.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Update, upgrade, and expand the performance of your FLIR moisture meter with a variety of probes, pins and other accessories.
Wall cavity pin probe for drywall, with 4" and 6" pins and impact plate. Hammer probe for subfloor and flooring, with 2" pins for subfloor and flooring.
6 Month Warranty
Specifications
  • Measurement TypeContact Pin
  • Height (in)2 5/64
  • Length (in)4 27/32
  • Depth (in)7 43/64
  • DescriptionWall Cavity Probe for MR-Series Moisture Meters