Flir DM66 TRMS Digital Multimeter with Non-Contact Voltage and Temperature
Representative image only.
Flir DM66 TRMS Digital Multimeter with Non-Contact Voltage and Temperature
Flir | Mfr: DM66| Item: 3572523
Diagnostic functions provide fast and effective troubleshooting
₹44,600.00INR / Each
for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹44,600.00INR / Each
Buy It Together
Current Item
Flir DM66 TRMS Digital Multimeter with Non-Contact Voltage and Temperature
Flir DM66 TRMS Digital Multimeter with Non-Contact Voltage and Temperature
3572523
₹44,600.00INR / Each
+
Flir TA60 Type K Thermocouple Probe with Adapter
2004842
₹9,200.00INR / Each
+ =
Total Price 81800.00 ADD 3 ITEMS TO CART
Product Description
Includes
Silicone test leads, alligator clips, integral probe holder, soft carrying case, and batteries.
Key Features
  • Automatically select the optimal measuring range with autoranging feature
  • Bright, large 6000-count 3.8” digital backlit LCD
  • Durable IP40 design for rugged applications
  • CAT III-600 V, CAT IV-300 V safety rating
  • 3 year warranty
More About this Item
These True RMS digital multimeters feature diagnostic measurements including 10 A AC/DC current, 600 V AC/DC voltage, and capacitance. Plus, you’ll work more efficiently with advanced features such as diode testing, continuity check, non-contact voltage detection, and min/max/average hold functions.
These multimeters are CAT III 600 V and CAT IV 300 V rated and can withstand a drop test up to 2 meters. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically on the large, backlit LCD with 5 reading per second update rate. Min/Max and average readings are recorded to capture variations.
Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
Measures ACV, DCV, Resistance, Continuity, Capacitance, Diode, DCA, ACA, Temperature, line Frequency, and non-contact Voltage (NCV)
Lo Z (low impedance) for ghost voltage protection
3 Year WarrantyConformite Europeenne