Flir CM275 Industrial Thermal Imaging AC/DC Clamp Meter with Datalogging, Wireless Connectivity and IGM™
Representative image only.
Flir CM275 Industrial Thermal Imaging AC/DC Clamp Meter with Datalogging, Wireless Connectivity and IGM™
Flir | Mfr: CM275| Item: 3572528
Visually identify electrical issues with this first thermal imaging clamp meter
for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Buy It Together
Current Item
Flir CM275 Industrial Thermal Imaging AC/DC Clamp Meter with Datalogging, Wireless Connectivity and IGM™
Flir CM275 Industrial Thermal Imaging AC/DC Clamp Meter with Datalogging, Wireless Connectivity and IGM™
3572528
₹153,000.00INR / Each
+
Flir TA55 AC Current Line Splitter
2684817
₹8,900.00INR / Each
+ =
Total Price 189900.00 ADD 3 ITEMS TO CART
Product Description
Includes
Silicone test leads, soft carrying case, and three AAA batteries.
Key Features
  • Work safely—scan a panel or cabinet for hazards using Infrared Guided Measurement (IGM™) without direct contact
  • Easily access difficult locations with the compact form-factor and narrow jaw
  • Laser and crosshair pinpoint problem locations through thermal images
  • Internally store measurements and thermal images and wirelessly share via Bluetooth®
  • 1 year warranty
More About this Item
The FLIR CM275 True RMS clamp meter with IGM is the first clamp meter equipped with a built-in thermal imager and true RMS, allowing you to quickly identify hot spots you can’t see with a standard clamp meter. Infrared Guided Measurement (IGM) technology visually guides you to the precise location of a potential electrical problem, identifying dangerous and unknown problem areas safely. Wirelessly connect your meter to FLIR Tools or FLIR InSite™ workflow management app for sharing data easily and quickly.