Fischer Technology MPOR Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
Representative image only.
Fischer Technology MPOR Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
Fischer Technology | Mfr: 605-097| Item: 5410222
Measures the thickness of coatings quickly and non-destructively using magnetic induction and Eddy current methods
Call For Price
Call For Price
Buy It Together
Current Item
Fischer Technology MPOR Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
Fischer Technology MPOR Dualscope Coating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current
5410222
Call For Price
+ =
Total Price 195000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Fe base, Al base, certified calibration foil, lanyard, carrying case, two AA batteries, and certificate of calibration supplied by the manufacturer (MP0R models also include Fischer Data Center Software with USB cable).
Key Features
  • 1 year warranty
More About this Item
These handheld coating thickness gauges have an integrated measurement probe for quick and easy results. The probe measures non-magnetic coatings like paint and enamel applied to ferrous substrates such as steel or iron using the magnetic induction method and/or measures non-conductive coatings applied to non-ferrous substrates such as aluminum and stainless steel, copper or brass, using the eddy current method. Gauges conform to ASTM D7091, B499, B244, DIN EN ISO 2178, 2360, 2808, SSPC-PA2 and more.
MPO Series features include display of mean value, standard deviation, min/max, number of measurements per block, and memory storage capability of up to 1000 measurements.
MPOR Series include a USB interface for bi-directional data transmission to Fischer Data Center Software allowing you download and store measurement data and create custom inspection reports. Gauges feature memory storage capability of up to 10,000 measurements.
1 Year Warranty
Specifications
  • Range0 to 80 mil
  • Measuring MethodEddy current, Magnetic induction
  • DescriptionCoating Thickness Gauge; 0-80 mils, Mag Induction/Eddy Current