Fischer Technology 604-187 Probe for inside measurements and recesses, 0 to 32 mils, Model FAI3.3-150, for Isoscope FMP
Representative image only.
Fischer Technology 604-187 Probe for inside measurements and recesses, 0 to 32 mils, Model FAI3.3-150, for Isoscope FMP
Fischer Technology | Mfr: 604-187| Item: 5410009
Fischer offers a wide range of probes for a variety of measurement tasks
₹322,000.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹322,000.00INR / Each
Buy It Together
Current Item
Fischer Technology 604-187 Probe for inside measurements and recesses, 0 to 32 mils, Model FAI3.3-150, for Isoscope FMP
Fischer Technology 604-187 Probe for inside measurements and recesses, 0 to 32 mils, Model FAI3.3-150, for Isoscope FMP
5410009
₹322,000.00INR / Each
+ =
Total Price 359400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • 6 month warranty
More About this Item
Choose the appropriate probe for your application.
6 Month Warranty
Specifications
  • Range0 to 32 mil (0 to 800 µm)
  • Accuracy±1 µm up to 200 µm; < 0.5% above 200 µm
  • Measuring MethodEddy current
  • Display240x160 pixels
  • ProbeInternal and recessed measurements
  • DescriptionProbe for inside measurements and recesses, 0 to 32 mils, Model FAI3.3-150, for Isoscope FMP