Extech TH30 USB Dual Temperature Datalogger
Representative image only.
Extech TH30 USB Dual Temperature Datalogger
Extech | Mfr: TH30| Item: 1800063
Measure ambient and external temperature simultanously
Call For Price
for an additional
₹40,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Extech TH30 USB Dual Temperature Datalogger
Extech TH30 USB Dual Temperature Datalogger
1800063
Call For Price
+ =
Total Price 40300.00 ADD 2 ITEMS TO CART
Product Description
Includes
Two CR2032 lithium batteries.
Key Features
  • Store up to 24,000 readings from each channel
  • Log data from every 30 seconds to every 2 hours
  • USB connection for easy data transfer
  • 1 year warranty
More About this Item
Model 18000-63 has two temperature channels—monitor external temperature with a stainless steel thermistor probe, and ambient temperature with the integral sensor. Order the optional probe (18000-78) to extend external temperature measurements 5-1/8" (13 cm)—probe 18000-76 is included.
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels2
  • Max Temperature (° F)158
  • Min Temperature (° F)-22
  • Max Temperature (° C)70
  • Min Temperature (° C)-30
  • Sampling Rate30 seconds to 2 hours
  • InterfaceUSB
  • AlarmStandard and delayed
  • Battery LifeApproximately 3 months
  • Battery Type2 CR2032 lithium (included)
  • DescriptionUSB Dual Temperature Datalogger