Extech TG20 Wire Tracer and Tone Generator
Representative image only.
Extech TG20 Wire Tracer and Tone Generator
Extech | Mfr: TG20| Item: 3572500
Quickly trace wires, check for breaks, and test phone lines
₹8,900.00INR / Each
₹8,900.00INR / Each
Product Description
Includes
wire tracer probe and transmitter, test lead set, and two 9 V alkaline batteries.
Key Features
  • Check electrical and telephone wiring with alligator clips and RJ11 connector
  • Insulated probe protects conductors
  • Easily interpret results using built-in speaker and colored LEDs
  • Trace cables or test for continuity, tip/ring, and telephone line condition
  • 1 year warranty
More About this Item
Use this wire tracer kit to quickly find cables, determine if a line is broken, and test telephone wires. Connect transmitter to wires using the alligator clips or insert the RJ11 plug directly into a telephone jack. Insulated probe protects cables under test from shorting. Built-in speaker with adjustable volume/sensitivity audibly alerts you when the correct cable is found. Green, yellow and red LEDs light up steadily, flicker, or dim and brighten in response to different wiring conditions and tests.
1 Year WarrantyConformite Europeenne
Specifications
  • DisplayLED
  • Battery2 9V (included)
  • DescriptionWire Tracer and Tone Generator