Extech SDL500 Datalogging Thermohygrometer with Sd Card Memory
Representative image only.
Extech SDL500 Datalogging Thermohygrometer with Sd Card Memory
Extech | Mfr: SDL500| Item: 3780371
Records data on an SD card in Excel® format for easy transfer to a PC for analysis
₹128,000.00INR / Each
for an additional
₹32,600.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹128,000.00INR / Each
Buy It Together
Current Item
Extech SDL500 Datalogging Thermohygrometer with Sd Card Memory
Extech SDL500 Datalogging Thermohygrometer with Sd Card Memory
3780371
₹128,000.00INR / Each
+ =
Total Price 160600.00 ADD 2 ITEMS TO CART
Product Description
Includes
Soft carrying case, humidity/temperature probe, 2 GB SD memory card and 6 AA batteries
Key Features
  • Four-function instrument measures humidity, dew point, wet-bulb and temperature
  • Datalogging capability up to 20,000 records using a 2G SD card
  • User programmable sampling rate from 1 to 3600 seconds
  • Manual store/recall of up to 99 records
  • Records readings with real date and time stamp
  • 1 year warranty
More About this Item
Four-function instrument measures humidity, dew point, wet-bulb and temperature.
Data is recorded onto a standard SD memory card, instead of a built-in memory. The advantage of the date being stored on an SD card is when the memory is full the SD card can be simply exchange out for unlimited data storage. The removable SD card is easy to transport and insert into a computer’s memory card slot or SD card reader for upload. The data is provided in Excel, allowing the user to easily analysis the information without the need for additional software. Data can also be streamed directly to a computer using the RS-232 port on the meter. Features include min/max, hold, temperature differential, offset adjustment, date and time stamp, auto power off and low battery indicator. Meter accepts an optional type-K or type-J thermocouple for measuring temperature only, for applications such as surface or high temperature measurements. User settable parameters include; date, time, sampling time, temperature unit of measure, and auto power off. Features include min/max, hold, date and time stamp, auto power off and low battery indicator.
1 Year Warranty
Specifications
  • Min RH Range (%)10
  • Max RH Range (%)95
  • Min Temperature (° F)-4
  • Max Temperature (° F)140
  • Min Probe Temperature (DegreeF)-58
  • Max Probe Temperature (DegreeF)2372
  • Display TypeLCD
  • OutputRS-232
  • Data LoggingYes
  • Battery6 AA (included)
  • DescriptionDatalogging Thermohygrometer with Sd Card Memory