Extech PQ34-12 Current Clamps, 1200 A, Set of 3

Representative image only.

Extech PQ34-12 Current Clamps, 1200 A, Set of 3

Extech | Mfr: PQ34-12| Item: 2004153

Measure current without power interruptions

₹225,000.00INR / Each

for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹225,000.00INR / Each

Buy It Together

Current Item

Extech PQ34-12 Current Clamps, 1200 A, set of 3
Extech PQ34-12 Current Clamps, 1200 A, set of 3
2004153

₹225,000.00INR / Each
+ =
Total Price 253000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Place around conductors without disconnecting and interrupting the power supply
  • 5 cm (2") jaw size
  • 1 year warranty
1 Year Warranty
Specifications
  • AC Max (amps)1200
  • Max Clamp Size2
  • DescriptionCurrent Clamps, 1200 A, Set of 3
  • Warranty1 Year
Reviews
Q&A