Extech MO257 Non-Evasive Moisture Meter with High Frequency Pinless Sensor
Representative image only.
Extech MO257 Non-Evasive Moisture Meter with High Frequency Pinless Sensor
Extech | Mfr: MO257| Item: 8653110
Monitor moisture in wood and other building materials with virtually no surface damage
₹67,700.00INR / Each
₹67,700.00INR / Each
Product Description
Includes
3 AA batteries and pouch case
Key Features
  • Relative pinless moisture reading for non-invasive measurement from 0.0 to 100.0 with 0.1 resolution
  • Multifunction backlit triple LCD display
  • Pinless measurement depth from 0.78" (20mm) to 1.6" (40mm) below the surface
  • High frequency sensing technology
  • Automatic Data Hold
  • Two adjustable alarm levels with audible beeper and visual (flashing “RISK” or “WET”) indicators
  • Automatic calibration (in dry air) when meter is powered on
  • Low battery indication
  • Auto Power off
  • 1 year warranty
1 Year WarrantyConformite Europeenne
Specifications
  • Moisture Range Reference Scale (Min)0
  • Moisture Range Reference Scale (Max)100
  • Display TypeDigital
  • AlarmYes
  • Battery3 AA (included)
  • DescriptionNon-Evasive Moisture Meter with High Frequency Pinless Sensor