Extech MN36 MULTIMETERMINIAUTORANGING
Representative image only.
Extech MN36 MULTIMETERMINIAUTORANGING
Extech | Mfr: MN36| Item: 2604636
Automatically recognizes proper measurement mode
₹11,400.00INR / Each
₹11,400.00INR / Each
Product Description
Includes
integrated test leads and two AAA batteries.
Key Features
  • Obtain True RMS readings up to 600 V AC/DC
  • Quickly detect voltage with built-in noncontact voltage (NCV) detector
  • Safely see in dark places—features LED flashlight
  • Compact size easily fits in your pocket or toolbox
  • 1 year warranty
More About this Item
The unique AutoSensing™ feature of the MN30 digital multimeter automatically changes modes for voltage or resistance readings. Additional features include a flashlight, audible continuity tester, overload and low-battery indication, and auto-off power—all with a CAT III-600 V safety rating.