Extech HD350 Anemometer and Differential Manometer with Pitot Tube
Representative image only.
Extech HD350 Anemometer and Differential Manometer with Pitot Tube
Extech | Mfr: HD350| Item: 1050901
Ideal in difficult-to-reach locations where a vane anemometer won''t fit.
₹139,000.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹139,000.00INR / Each
Buy It Together
Current Item
Extech HD350 Anemometer and Differential Manometer with Pitot Tube
Extech HD350 Anemometer and Differential Manometer with Pitot Tube
1050901
₹139,000.00INR / Each
+ =
Total Price 176400.00 ADD 2 ITEMS TO CART
Product Description
Includes
What''s included: Windows® compatible software with cable, 9V battery, pitot tube with two 33.5” (85cm) connection hoses, 100V-240V universal AC adaptor, and hard carrying case.
Key Features
  • Simultaneous display of Pressure, Air Velocity or Air Flow plus Temperature
  • ± 0.7252psi range
  • 5 selectable units of pressure measurement
  • Max/Min/Avg recording and Relative time stamp
  • Data hold and Auto power off functions
  • Large LCD display with backlighting
  • Zero function for offset correction or measurement
  • Stoer/Recall up to 99 readings in each mode
  • USB port includes software
  • 3 year warranty
3 Year WarrantyConformite Europeenne
Specifications
  • DescriptionAnemometer and Differential Manometer with Pitot Tube