Extech DV30 AC Voltage Detector Non-Contact Adjustbl
Representative image only.
Extech DV30 AC Voltage Detector Non-Contact Adjustbl
Extech | Mfr: DV30| Item: 2002120
₹6,900.00INR / Each
₹6,900.00INR / Each
Product Description
Includes
pocket clip and two LR44 button batteries.
Key Features
  • 1 year warranty
More About this Item
Sensitivity adjusts to pinpoint voltage source
  • Non-contact detection of AC voltage from 12VAC to 600VAC
  • Sensitivity adjustment increases or reduces sensor trigger threshold
  • Audible and visible voltage indication
  • Self test function
  • Small size with pocket clip
  • Dimensions: 6.1 x 1.1 x 1" (155 x 28 x 25mm); Weight: 1.5 oz (42g)
1 Year Warranty
Specifications
  • DescriptionAC Voltage Detector Non-Contact Adjustbl