Extech DT4208SD 12 Channel Datalogging Thermometer
Representative image only.
Extech DT4208SD 12 Channel Datalogging Thermometer
Extech | Mfr: TM500| Item: 3975905
12-Channel temperature data logging with 6 Thermocouple types (J, K, E, T, R,S)
₹339,000.00INR / Each
₹339,000.00INR / Each
Product Description
Includes
twelve type K beaded wire temperature probes, 2 G SD card, hand carrying case, and eight AA batteries.
Key Features
  • Simultaneously displays CH1 to CH8 or CH9 to CH12
  • Offset adjustment used for zero function or to make relative measurements
  • Stores 99 readings manually and 20M readings via 2GB SD card
  • Records readings with date and time stamp
  • User-programmable sampling rate: 1 to 3600 seconds
  • Min/Max and Data Hold functions
  • Auto power off with disable function
  • 1 year warranty
More About this Item
This 12-channel meter has automatic temperature compensation and is designed to work with the six most popular thermocouple types: J, K, T, E, S, and R. Offers data logging capability up to 20,000 records using a 2 G SD card, with user-programmable sampling rate from one second up to one hour. The meter displays 6 of the 12 channels at a given time (Ch 1 to 8 or Ch 9 to 12).
Data is recorded either on the internal memory up to 99 readings or onto a standard SD memory card, offering unlimited data storage. The data on the SD card is provided in an Excel format. Data can also be streamed directly to a computer using the RS-232 port on the meter.
Additional meter features include a large 2-1⁄2" backlit display, Min/Max, Hold, offset adjustment, loop data recording, sampling interval, date-and-time stamp, selectable units of measure, period or comma decimal point division, low-battery indication, and auto power-off. The unit operates on either battery or AC power.
1 Year Warranty
Specifications
  • Width (InchShort)5
  • Min Temperature (° F)-148
  • Height (InchShort)8 55/64
  • Max Temperature (° F)2372
  • Min Temperature (° C)-100
  • Max Temperature (° C)1300
  • Description12 Channel Datalogging Thermometer