Extech CT20 Remote and Local Continuity Tester Pro
Representative image only.
Extech CT20 Remote and Local Continuity Tester Pro
Extech | Mfr: CT20| Item: 2604633
One person operation for wire and cable testing
₹11,400.00INR / Each
₹11,400.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
The ideal device for checking the continuity of wiring, fuses, switches, relays, etc. The bright flashing LED light and loud pulsating beeper provide positive continuity checks in a noisy working environment. The pocket sized device is light enough to be suspended by the attached alligator clip connections. No need to prop it up. Allows hands-free operation.The Continuity Tester Pro can also identify wire pairs or multiple cables remotely by using the external probe. The probe's bi-color LED light will flash green or red depending on polarity. Can be used to label and identify up to three cables with only one trip to the end location -cuts down on multiple trips back and forth.
1 Year Warranty
Specifications
  • DescriptionRemote and Local Continuity Tester Pro