Extech CL207 Clorine Calibration Standard, 1 ppm, 3 x 20 mL Ampules

Representative image only.

Extech CL207 Clorine Calibration Standard, 1 ppm, 3 x 20 mL Ampules

Extech | Mfr: CL207| Item: 9500195

Ensure accuracy of your chlorine meters

₹16,000.00INR / Each

₹16,000.00INR / Each
Product Description

Key Features

  • 1 ppm concentration
  • Three 20 mL ampules
  • 6 month warranty

More About this Item

Calibrate your chlorine meters to ensure your measurements are accurate.

Conformite Europeenne6 Month Warranty
Specifications
  • Compatible WithChlorine meters
  • DescriptionClorine Calibration Standard, 1 ppm, 3 x 20 mL Ampules
  • Warranty6 month
Reviews
Q&A