Extech CB10-Kit Electrical Trouble Shooting Kit
Representative image only.
Extech CB10-Kit Electrical Trouble Shooting Kit
Extech | Mfr: CB10-Kit| Item: 2003736
Troubleshooting kit performs up to five functions
₹34,300.00INR / Each
for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹34,300.00INR / Each
Buy It Together
Current Item
Extech CB10-Kit Electrical Trouble Shooting Kit
Extech CB10-Kit Electrical Trouble Shooting Kit
2003736
₹34,300.00INR / Each
+ =
Total Price 62300.00 ADD 2 ITEMS TO CART
Product Description
Includes
Two 9 V batteries (one each for models CB10 and CT20), and two AA batteries (model DV20)
Key Features
  • CB10 Circuit Breaker Finder
  • Quickly locate circuit breakers with audible and visual alert
  • Variable sensitivity adjustment to pinpoint correct circuit breaker
  • LED indicators determine if outlet is correctly wired, and identifies six fault conditions
  • Push button GFCI test
  • UL listed and CE approved
  • CT20 Continuity Tester/Wire Tracer
  • Continuity testing via audible and visual alarms
  • Remote probe with flashing (red/green) LED’s - trace up to three cables
  • Easily clips on and hangs from the cable(s) under test without support
  • CE approved
  • DV20 AC Voltage Detector/Flashlight
  • Non-contact detection of AC voltage from 100 VAC to 600 VAC
  • Rugged, double molded housing
  • Built-in bright LED flashlight with ON/OFF button
  • UL listed, CE approved - CAT lll 600 V rated
  • 1 year warranty
More About this Item
Save time and money conducting electrical circuit testing and preventive maintenance checks ensuring all your systems are operating optimally and efficiently with this electrical troubleshooting kit. The kit combines a circuit breaker finder with outlet/GFCI tester, continuity checker/wire tracer, and AC voltage detector with flashlight, in one convenient kit.
1 Year Warranty
Specifications
  • DescriptionElectrical Trouble Shooting Kit