Extech AN300-C Airflow Cone and Funnel Adapters
Representative image only.
Extech AN300-C Airflow Cone and Funnel Adapters
Extech | Mfr: AN300-C| Item: 1000072
Provides quick and easy means of airflow measurement for common duct sizes
₹80,400.00INR / Each
₹80,400.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Optional AN300-C airflow cone and funnel adapter kit provides a quick and easy means of airflow measurement for common duct sizes: 8-1/4" (210 mm) round and 13-5/8" x 13-5/8" (346 x 346 mm) square.
6 Month WarrantyConformite Europeenne
Specifications
  • DescriptionAirflow Cone and Funnel Adapters