Extech 480172 3-Prong 120 VAC Line Separator

Representative image only.

Extech 480172 3-Prong 120 VAC Line Separator

Extech | Mfr: 480172| Item: 2684254

₹5,300.00INR / Each

₹5,300.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

AC line separator provides temporary separation of conductors for current measurement with clamp-type meters. Simply plug the separator into a grounded 120 VAC receptacle, plug in your equipment and turn it on, then measure.

1 Year Warranty
Specifications
  • Safety RatingCAT III 300V, EN 61010-1
  • Description3-Prong 120 VAC Line Separator
  • Warranty1 Year
Reviews
Q&A