Extech 461937 Tachometer Reflective Tape; 10/Pk
Representative image only.
Extech 461937 Tachometer Reflective Tape; 10/Pk
Extech | Mfr: 461937| Item: 8770262
Enhance measurement accuracy and performance of your tachometer
₹12,100.00INR / Pkg of 10
₹12,100.00INR / Pkg of 10
Product Description
Key Features
  • Each strip measures 23"L x 1/4"W
  • Pack of 10
More About this Item
Tachometer optical reflective tape provides good reflective signal for any optical (laser or standard) tachometer.
Specifications
  • Compatible WithNoncontact tachometers
  • Length (InchShort)23
  • Width (InchShort)1/4
  • Width (mm)6
  • Length (mm)584
  • Quantity10
  • DescriptionTachometer Reflective Tape; 10/Pk