Extech 407766-NIST Dual Calibrator with NIST-Traceable Certificate, 94/114 dB
Representative image only.
Extech 407766-NIST Dual Calibrator with NIST-Traceable Certificate, 94/114 dB
Extech | Mfr: 407766-NIST| Item: 0161861
₹194,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹194,000.00INR / Each
Product Description
Includes
calibration tool, case, and two 9 V batteries
Key Features
  • Calibrate sound meters in the field
  • 1 year warranty
More About this Item
Lightweight and portable calibrator performs daily calibrations of your sound meter prior to use — ideal for ensuring precise readings in the field or lab. Accommodate sound meters with 1/2" dia microphones. Simply place calibrator on microphone, turn on, and adjust your sound meter.
1 Year Warranty
Specifications
  • DescriptionDual Calibrator with NIST-Traceable Certificate, 94/114 db