Extech 39272 Adjustable Angle Thermometer
Representative image only.
Extech 39272 Adjustable Angle Thermometer
Extech | Mfr: 39272| Item: 9038200
Convenient pen-like design fits in your pocket
₹16,100.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹16,100.00INR / Each
Buy It Together
Current Item
Extech 39272 Adjustable Angle Thermometer
Extech 39272 Adjustable Angle Thermometer
9038200
₹16,100.00INR / Each
+ =
Total Price 34800.00 ADD 2 ITEMS TO CART
Product Description
Includes
One AAA battery and wrist strap.
Key Features
  • Hinged design allows probe to fold away
  • 1 year warranty
More About this Item
This thermometer takes fast and accurate measurements anywhere. Tapered probe can be used in solids, semisolids, liquids and gases. Large ½” high display provides great visibility. Fold up 4.5”L (114mm) probe adjusts from 0° to 180° for best measurement and viewing angle. Features data hold and min/max functions. Auto shut-off after one hour of non-use to conserve battery life. What’s included: One AAA battery and wrist strap
1 Year Warranty
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels1
  • Min Temperature (° F)-58
  • Max Temperature (° F)572
  • Min Temperature (° C)-50
  • Max Temperature (° C)300
  • Resolution0.1°
  • Display Type0.5" Display, 3.5-digit LCD
  • Height (in)2
  • Depth (in)3/4
  • No. of Probe(s) Included1
  • Probe Length (in)4 1/2
  • Battery1 AAA (included)
  • DescriptionAdjustable Angle Thermometer