Extech 382357 Clamp-On Ground Resistance Tester
Representative image only.
Extech 382357 Clamp-On Ground Resistance Tester
Extech | Mfr: 382357| Item: 2000314
₹370,000.00INR / Each
for an additional
₹47,100.00INR / Each
for an additional
₹34,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹370,000.00INR / Each
Buy It Together
Current Item
Extech 382357 Clamp-On Ground Resistance Tester
Extech 382357 Clamp-On Ground Resistance Tester
2000314
₹370,000.00INR / Each
+ =
Total Price 404900.00 ADD 2 ITEMS TO CART
Product Description
Includes
9V battery, Resistance check plate and carrying case.
Key Features
  • 1 year warranty
More About this Item
Enables non-contact measurements of ground conductors without need for auxiliary ground spikes
FEATURES
  • Autoranging ground resistance measurements from 0.03 O to 1500 O with 0.02 O resolution
  • True RMS AC Leakage current range of 0.2mA to 15.00A
  • Programmable datalogging with 116 data points
  • Programmable Hi/Lo alarm
  • 0.9” (23mm) Jaw size for large ground rods
  • Simplifies ground resistance measurements on multiple point ground systems
  • Verification resistors confirm accuracy of meter
  • Automatic Self Calibration when powered on
  • 9999 Count LCD display with Data Hold
  • Electrical Noise Detection feature prevents inaccurate readings
  • CAT III-300V, CAT II-600V; CE
  • One year warranty
1 Year Warranty
Specifications
  • DescriptionClamp-On Ground Resistance Tester