Expo Engineering HDD-55/240V Heated Drum Dolly, Fits 55 gal, 550° F, 240 V

Representative image only.

Expo Engineering HDD-55/240V Heated Drum Dolly, Fits 55 gal, 550° F, 240 V

Expo Engineering | Mfr: HDD-55/240V| Item: 0950030

Mobile drum heater provides operator flexibility

₹218,000.00INR / Each

₹218,000.00INR / Each
Product Description

Key Features

  • Dolly supports up to 900 pound capacity
  • 1 year warranty

More About this Item

Use this drum heated dolly to warm your nonflammable, nonhazardous material contained within 55-gallon metal drums. The built-in infinite control (0 to 100% adjustable) features on/off as well as heat pulsing action. Drums rest on a spring-loaded 15 1/4” square by 1/4” thick aluminum heat platen, and rolls on 3"-diameter steel swivel casters that feature two-wheel locking capability. Measure the temperature of drum contents with an external temperature measurement device (not included) for more precise temperature control.

1 Year Warranty
Specifications
  • Drum MaterialMetal
  • Drum Size55 gallons
  • Max Temperature (° F)550
  • Max Temperature (° C)288
  • Min TemperatureAmbient;Ambient
  • Plug TypeUS Standard
  • Power (VAC)240
  • Power (amps)8
  • Power (watts)1920
  • Diameter (in)22.3 in
  • DescriptionHeated Drum DOlly, Fits 55gal, 550F, 240 V
  • Warranty1 year
Reviews
Q&A