Excel Scientific ThinSeal Microplate Film, 25-µm Thick, Sterile; 100/pk, Sterile
Representative image only.
Excel Scientific ThinSeal Microplate Film, 25-µm Thick, Sterile; 100/pk, Sterile
Excel Scientific | Mfr: STR-THIN-PLT| Item: 1302462
Prevents evaporation and protects samples during processing, storage, and shipment
₹24,300.00INR / Pkg of 100
₹24,300.00INR / Pkg of 100
Product Description
Key Features
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from -40 to 120°C
  • 3 year warranty
More About this Item
ThinSeal™ films are just 25-µm thick, clearer and marginally piercable with a single-channel pipette or probe for sample recovery. Measure 79.4 x 141 mm.
3 Year Warranty
Specifications
  • SterileYes
  • Plate TypeSealPlate® ThinSeal™ films
  • Min Temperature (° C)-40
  • Max Temperature (° C)120
  • MaterialPolyester
  • Width (mm)79.4
  • Length (mm)141
  • DescriptionThinSeal Microplate Film, 25-μm Thick, Sterile; 100/pk, Sterile