Excel Scientific Thermal Cycling Film, Polyolefin, 50um, PCR; 100/Pk
Representative image only.
Excel Scientific Thermal Cycling Film, Polyolefin, 50um, PCR; 100/Pk
Excel Scientific | Mfr: TSS-RTQ-100| Item: 1302444
Encapsulated silicone adhesive binds only when pressure is applied – will not stick to gloves
₹48,800.00INR / Case of 100
₹48,800.00INR / Case of 100
Product Description
Includes
100 films.
Key Features
  • Ideal for real-time qPCR, storage, and protein crystallization
More About this Item
These sealing films are made of a unique, 50µm polyolefin film that utilizes an encapsulated silicone adhesive that is non-tacky until pressed in place on microplates — eliminates sticking to gloves. Adhesive only seals around wells, keeping the area over the wells encapsulated and inert. Film fits within the raised area of 96-well plates, preventing loss of seal. White tabs on either end of film provide easier positioning on the plate.
Heat resistant films offer a high optical clarity with minimal autofluorescence. Film is chemically inert (except at extreme pH) and offers DMSO resistance for high through-put screening. Films are certified Dnase-, Rnase-, and nucleic acid-free.
Specifications
  • Plate ColorClear
  • Min Temperature (° C)-70
  • Max Temperature (° C)100
  • MaterialPolyolefin
  • Width (mm)133.4
  • Length (mm)76.2
  • DescriptionThermal Cycling Film, Polyolefin, 50um, PCR; 100/Pk