Excel Scientific Films, Sterile; 100/Pk
Representative image only.
Excel Scientific Films, Sterile; 100/Pk
Excel Scientific | Mfr: STR-SEAL-PLT| Item: 1302402
Prevents evaporation and protects sample during processing, storage, and shipment
₹18,600.00INR / Pkg of 100
₹18,600.00INR / Pkg of 100
Product Description
Key Features
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
More About this Item
SealPlate Films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover. Secure sealing of all wells eliminates edge effects in sensitive ELISA assays. Films can also be used with tissue culture plates for shortterm storage, incubation, and containment of biohazards. These 50-ìm thick polyester films are non-pierceable. Each measures 79.4 x 103.2 mm.
Sterile
Specifications
  • SterileYes
  • Plate TypeSealPlate film
  • Min Temperature (° C)-40
  • Max Temperature (° C)120
  • MaterialPolyester
  • Width (mm)79.4
  • Length (mm)146.1
  • DescriptionFilms, Sterile; 100/Pk