Erlab HALO Chemplus Laboratory Grade Air Purification Station; 80 to 240 VAC

Representative image only.

Erlab HALO Chemplus Laboratory Grade Air Purification Station; 80 to 240 VAC

Erlab | Mfr: HALOCHEMPLUS| Item: 3352031

Provides healthier, cleaner air in your laboratory

Call For Price

Call For Price

Buy It Together

Current Item

Erlab HALO Chemplus  Laboratory Grade Air Purification Station; 80 to 240 VAC
Erlab HALO Chemplus Laboratory Grade Air Purification Station; 80 to 240 VAC
3352031

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Includes

Four eye bolts to mount unit to ceiling.

Key Features

  • Healthier air with considerable savings
  • Simple to install and use
  • Easy access panel for quick filter changes
  • BACnet capable
  • Network the units to monitor remotely
  • 1 year warranty

More About this Item

Halo 32 1C Laboratory Grade Air Purification Station is easily installed in the ceiling, but does not need to be tied in to existing HVAC. As a standalone unit, it is designed to effectively clean 250-300 sq. ft of space, and it can be placed anywhere that would be most effective for you, You can use multiple units to cover larger areas. It works to purify the air from fugitive emissions present in the lab, while also providing real-time communication and 24/7 data with the lab manager and users of the air pollution levels.

Each Halo comes with Erlab’s state-of-the-art filters either for VOC capture, Chemplus for capture of a broad range of chemicals from solvents (organic and in-organic), acids and bases, or Formaldehyde capture. The laboratory-grade molecular filtration system contains Neutrodine, a unique and powerful activated carbon mixture combined with a pre-filter that captures a large variety of airborne fumes and odors, which left untreated, could represent a potential health hazard. In addition to purifying the lab air, the Halo is also effective in the reduction of the air exchange rates per hour (ACH) needed to reduce possible concentration of pollutants.

Comes with Smart Technology™, an exclusive set of tools that consists of Smart-Light Communication™, a soft band of light assures the occupants of the lab that the air is clean. When the LED lights slowly pulse, HALO is letting you know it is sensing and capturing fugitive emissions. You can easily check the status, change the setting, or check for routine maintenance via your smartphone, tablet or PC.

1 Year Warranty
Specifications
  • Exterior Width (in)35.125
  • Exterior Height (in)23.3125
  • Exterior Depth (in)10.25
  • Power (VAC)85 to 240
  • Power (Hz)50/60
  • Exhaust Volume (CFM)135
  • Filter TypeSolvents and acids
  • DescriptionChemplus Laboratory Grade Air Purification Station; 80 to 240 VAC
  • Warranty1 Year
Reviews
Q&A