Erie Scientific Clipped-Corner Slides, Superfrost, 144/Pk
Representative image only.
Erie Scientific Clipped-Corner Slides, Superfrost, 144/Pk
Erie Scientific | Mfr: 4951-4-001| Item: 4852005
Corners are clipped for easy handing
₹16,300.00INR / Pkg of 144
₹16,300.00INR / Pkg of 144
Product Description
Key Features
  • Clipped corners keep sample away from the edges of the slide—ideal for making smears
More About this Item
Drastically reduce the chances of cutting your finger or tearing a glove when you use these slides. Slides measure 75 x 25 mm and are 1-mm thick.
Superfrost slides provide a chemically-resistant, opaque white writing surface for easy labeling.
Specifications
  • Slide TypeSuperfrost
  • Size75 x 25 mm
  • DescriptionClipped-Corner Slides, Superfrost, 144/PK