Eppendorf Filter Tips, 2-20 µL, 96 per rack
Representative image only.
Eppendorf Filter Tips, 2-20 µL, 96 per rack
Eppendorf | Mfr: 0030078535| Item: 1888824
Protect your pipettor and prevent cross-contamination
₹112,000.00INR / Case of 960
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹112,000.00INR / Case of 960
Buy It Together
Current Item
Eppendorf Filter Tips, 2-20 µL, 96 per rack
Eppendorf Filter Tips, 2-20 µL, 96 per rack
1888824
₹112,000.00INR / Case of 960
+ + =
Total Price 424500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Integrated hydrophobic filters do not contain "self-sealing" additives. All filter tips are sterile and PCR clean (free of DNA, DNase, RNase, and PCR inhibitors). Packaged in racks of 96 tips.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (µL)20
  • Min Volume (µL)2
  • Min Volume (mL)0.002
  • Max Volume (mL)0.02
  • Tip Volume (mL)0.02
  • Min Dispensing (mL)0.0002
  • Max Dispensing (mL)0.02
  • Color CodeYellow
  • Package Of10 racks; 96 tips per rack
  • DescriptionFilterTips,2to20uL,96perrack