Eppendorf Filter Tips, 2-100 µL, 96 per rack
Representative image only.
Eppendorf Filter Tips, 2-100 µL, 96 per rack
Eppendorf | Mfr: 0030078543| Item: 1888826
Protect your pipettor and prevent cross-contamination
₹108,000.00INR / Case of 960
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹108,000.00INR / Case of 960
Buy It Together
Current Item
Eppendorf Filter Tips, 2-100 µL, 96 per rack
Eppendorf Filter Tips, 2-100 µL, 96 per rack
1888826
₹108,000.00INR / Case of 960
+ + =
Total Price 420500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Integrated hydrophobic filters do not contain "self-sealing" additives. All filter tips are sterile and PCR clean (free of DNA, DNase, RNase, and PCR inhibitors). Packaged in racks of 96 tips.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (µL)100
  • Min Volume (µL)2
  • Min Volume (mL)0.002
  • Max Volume (mL)0.1
  • Tip Volume (mL)0.1
  • Min Dispensing (mL)0.0002
  • Max Dispensing (mL)0.1
  • Color CodeYellow
  • Package Of10 racks; 96 tips per rack
  • DescriptionFilterTips,2to100uL,96perrack