Eppendorf Filter Tips, 0.1-10 µL, small
Representative image only.
Eppendorf Filter Tips, 0.1-10 µL, small
Eppendorf | Mfr: 0030078500| Item: 1888818
Protect your pipettor and prevent cross-contamination
₹114,000.00INR / Case of 10
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹114,000.00INR / Case of 10
Buy It Together
Current Item
Eppendorf Filter Tips, 0.1-10 µL, small
Eppendorf Filter Tips, 0.1-10 µL, small
1888818
₹114,000.00INR / Case of 10
+ + =
Total Price 426500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Integrated hydrophobic filters do not contain "self-sealing" additives. All filter tips are sterile and PCR clean (free of DNA, DNase, RNase, and PCR inhibitors). Packaged in racks of 96 tips.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • SterileYes
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Min Volume (mL)0.0001
  • Max Volume (mL)0.01
  • Tip Volume (mL)0.01
  • Min Dispensing (mL)0.0001
  • Max Dispensing (mL)0.01
  • Color CodeGray
  • Package Of10 racks; 96 tips per rack
  • DescriptionFilterTips,0.1to10µL,small