Eppendorf epTIPS Sterile , Individually Wrapped Singles, 2 to 200 uL, 1000/CS
Representative image only.
Eppendorf epTIPS Sterile , Individually Wrapped Singles, 2 to 200 uL, 1000/CS
Eppendorf | Mfr: 022492209| Item: 1888952
High standards of purity ideally suited for critical laboratory applications
₹106,000.00INR / Case of 1000
₹106,000.00INR / Case of 1000
Product Description
Includes
certificate of purity/sterility.
Key Features
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose sterile tips for microbiology and cell technology applications
More About this Item
Pipette tips sterilized by beta radiation maintain purity greater than those exposed to ethylene oxide, which is a gas absorbed by plastics.
Specifications
  • SterileYes
  • Max Volume (µL)200
  • Min Volume (µL)2
  • Min Volume (mL)0.002
  • Max Volume (mL)0.2
  • Tip Volume (mL)0.2
  • Min Dispensing (mL)0.002
  • Max Dispensing (mL)0.2
  • Package OfIndividually Wrapped Singles; 1000/CS
  • DescriptionepTIPS Sterile , Individually Wrapped Singles, 2 to 200 uL, 1000/CS