Eppendorf epTIPS Single Use Racks 2 to 200 uL 480/Pk
Representative image only.
Eppendorf epTIPS Single Use Racks 2 to 200 uL 480/Pk
Eppendorf | Mfr: 0030075234| Item: 1888808
High standards of purity ideally suited for critical laboratory applications
₹66,000.00INR / Case of 480
₹66,000.00INR / Case of 480
Product Description
Includes
certificate of purity/sterility.
Key Features
  • Pyrogen-free (endotoxin-free) according to LAL test
  • Sterilized by beta radiation to SAL 10-6, avoiding contamination by ethylene oxide
  • Tested and certified by an independent laboratory
  • Choose Biopur tips for molecular and microbiology, cell technology, quality control, and forensic applications
More About this Item
Pipette tips sterilized by beta radiation maintain purity greater than those exposed to ethylene oxide, which is a gas absorbed by plastics.
Additional certifications for Biopur tips include DNA-free, DNase-free, RNase-free, PCR inhibitor-free, and ATP-free.
Specifications
  • SterileNo
  • Max Volume (µL)200
  • Min Volume (µL)2
  • Min Volume (mL)0.002
  • Max Volume (mL)0.2
  • Tip Volume (mL)0.2
  • Min Dispensing (mL)0.002
  • Max Dispensing (mL)0.2
  • Package OfIndividually Wrapped Single Use Racks; 480/Pk
  • DescriptionepTIPS Single Use Racks 2 to 200 uL 480/Pk