EPC3020 Cubitainer, LDPE, 4 Liter; 12/Cs
Representative image only.
EPC3020 Cubitainer, LDPE, 4 Liter; 12/Cs
| Mfr: EPC3020| Item: 3520481
Collapsible containers nest for efficient storage
₹16,400.00INR / Case of 12
₹16,400.00INR / Case of 12
Buy It Together
Current Item
EPC3020 Cubitainer, LDPE, 4 Liter; 12/Cs
EPC3020 Cubitainer, LDPE, 4 Liter; 12/Cs
3520481
₹16,400.00INR / Case of 12
+ =
Total Price 19700.00 ADD 2 ITEMS TO CART
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • 3 month warranty
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Natural LDPE containers come assembled with LDPE-lined size 38 caps.
3 Month Warranty
Specifications
  • Capacity (mL)4000
  • Capacity (L)4
  • Capacity (oz)128
  • Capacity (Gal)1
  • MaterialLow Density Polyethyene (LDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, LDPE, 4 Liter; 12/Cs