Environmental Express Unassembled Filter Funnel, membrane not included, 400/pk
Representative image only.
Environmental Express Unassembled Filter Funnel, membrane not included, 400/pk
Environmental Express | Item: 3520055
Prevent sample-to-sample contamination to ensure accurate alpha particle spectra
₹71,400.00INR / Pkg of 1
₹71,400.00INR / Pkg of 1
Product Description
Key Features
  • Polypropylene construction withstands exposure to acids and bases
  • Choose convenient preassembled filter units or separate funnels and filters to accommodate any protocol
More About this Item
Total funnel capacity is 68 mL and filter diameter is 25 mm. Funnel stem fits a standard rubber stopper hole. Easily remove the filter to analyze filtrate.
Unassembled filter funnels include a top cap and filter support. Filters sold separately.
Specifications
  • Membrane Material
  • Housing MaterialPolypropylene (PP)
  • Sample Volume (mL)68
  • DescriptionUnassembled Filter Funnel, membrane not included, 400/pk