Environmental Express UltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK
Representative image only.
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK
Environmental Express | Item: 3521038
Convenience of disposable, unbreakable funnels for oil and grease analysis
₹28,800.00INR / Pkg of 20
₹28,800.00INR / Pkg of 20
Buy It Together
Current Item
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK
3521038
₹28,800.00INR / Pkg of 20
+ =
Total Price 48100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose between UltraPrep and UltraFlow extraction funnels
  • Available in 47-mm; Pack of 20
  • 3 month warranty
More About this Item
StepSaver streamlines the extraction process by allowing the analyst to collect the solvent with eluted oil and grease directly into a collection flask. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. It is ready to be evaporated without a separate drying step.
UltraPrep disposable polypropylene filter funnels house 47mm UltraPrep SPE filter disks. Disposable funnels are designed to fit the Environmental Express StepSaver Oil and Grease Extraction System.
UltraPrep Premium Extraction Disks follow Method 1664 for solid phase extraction of oil & grease but eliminate the need for conditioning with a polar solvent. See UltraPrep Filters, GUP047MM.
Polypropylene, disposable funnel housing has over 250mL total capacity. Tests show no measurable extractable levels in the polypropylene housings.
20 UltraFlow filter funnels, 20 per pack.
3 Month Warranty
Specifications
  • Filter Diameter (mm)47
  • Body MaterialPolypropylene
  • Funnel Size (mL)250
  • DescriptionUltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK