Environmental Express UltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK
Representative image only.
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK
Environmental Express | Item: 3521039
Convenience of disposable, unbreakable funnels for oil and grease analysis
₹47,100.00INR / Pkg of 20
₹47,100.00INR / Pkg of 20
Buy It Together
Current Item
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK
Environmental Express UltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK
3521039
₹47,100.00INR / Pkg of 20
+ =
Total Price 66400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose between UltraPrep and UltraFlow extraction funnels
  • Available in 47-mm; Pack of 20
  • 3 month warranty
More About this Item
StepSaver streamlines the extraction process by allowing the analyst to collect the solvent with eluted oil and grease directly into a collection flask. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. It is ready to be evaporated without a separate drying step.
3 Month Warranty
Specifications
  • Filter Diameter (mm)47
  • Body MaterialPolypropylene
  • Funnel Size (mL)250
  • DescriptionUltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK