Environmental Express Tuff-Tip, Semi-Domed 1" In-Line or Submersible pH Electrode, No ATC, BNC
Representative image only.
Environmental Express Tuff-Tip, Semi-Domed 1" In-Line or Submersible pH Electrode, No ATC, BNC
Environmental Express | Item: 2702020
Less chance of breakage
₹40,800.00INR / Each
₹40,800.00INR / Each
Buy It Together
Current Item
Environmental Express Tuff-Tip, Semi-Domed 1" In-Line or Submersible pH Electrode, No ATC, BNC
Environmental Express Tuff-Tip, Semi-Domed 1" In-Line or Submersible pH Electrode, No ATC, BNC
2702020
₹40,800.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ +
Cole-Parmer Epoxy pH Electrode BNC
0599223
₹23,200.00INR / Each
+ =
Total Price 248900.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose chemical-resistant Ryton® PPS or graphite-tipped electrodes
  • 1 year warranty
More About this Item
Don't worry about bulb breakage— unique design helps protect bulbs
– Eliminate electrical noise with graphite-tipped models
– Ideal for viscous or severe applications
– Reduce contamination and clogging with uniform pore size
Keep a Spare1 Year Warranty
Specifications
  • Electrode Body MaterialKynar®
  • Reference JunctionCeramic
  • Max Temperature (° C)110
  • ATC ConnectorNone
  • ATC ElementNone
  • ConnectorBNC
  • DescriptionTuff-Tip, Semi-Domed 1" In-Line or Submersible pH Electrode, No ATC, BNC