Environmental Express StableWeigh Filling Station, Single Place
Representative image only.
Environmental Express StableWeigh Filling Station, Single Place
Environmental Express | Item: 3521108
StableWeigh Kits provide a time-saving alternative to traditional TDS methods
₹320,000.00INR / Each
₹320,000.00INR / Each
Buy It Together
Current Item
Environmental Express StableWeigh Filling Station, Single Place
Environmental Express StableWeigh Filling Station, Single Place
3521108
₹320,000.00INR / Each
+ +
Environmental Express StableWeigh TDS Vessels, 100/pk
3521109
₹59,500.00INR / Pkg of 100
=
Total Price 379500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007
  • Save time — cools faster, requires fewer drying and weighing cycles
  • Disposable to eliminate cross-contamination
More About this Item
These weighing vessels reduce preparation time, perfect for busy labs analyzing a lot of samples. Each vessel complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007. Each vessel is treated to conform with ASTM D5907 Standard Test Methods for Filterable Matter (Total Dissolved Solids) and Nonfilterable Matter (Total Suspended Solids) in Water, then individually printed with the final weight. The material is lightweight, inert to water or wastewater components, and heat resistant. Vessels cool quickly decreasing wait times between heating and weighing.
Specifications
  • DescriptionStableWeigh Filling Station, Single Place